missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Macrophage mannose receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Macrophage mannose receptor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6378-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002429.1 | |
Macrophage mannose receptor 1 Protein | |
Research Use Only | |
MRC1 | |
Recombinant Protein | |
Yeast | |
RTSLCFKLYA KGKHEKKTWF ESRDFCRALG GDLASINNKE EQQTIWRLIT ASGSYHKLFW LGLTYGSPSE GFTWSDGSPV SYENWAYGEP NNYQNVEYCG ELKGDPTMSW NDINCEHLNN WICQIQKGQT PKPEPTPAPQ DNPPVTEDGW VIYKDYQYYF SKEKETMDNA RAFCKRNFGD LVSIQSESEK KFLWKYVNRN DAQSAYFIGL LISLDKKFAW MDGSKVDYVS WATGEPNFAN EDENCVTMYS NSGFWNDINC GYPNAFICQR HNSSINATTV MPTMPSVPSG CKEGWNFYSN KCFKIFGFME EERKNWQEAR KACIGFGGNL VSIQNEKEQA FLTYHMKDST FSAWTGLNDV NSEHTFLWTD GRGVHYTNWG KGYPGGRRSS LSYEDADCVV IIGGASNEAG KWMDDTCDSK RGYICQTRSD PSLTNPPATI QTDGFVKYGK SSYSLMRQKF QWHEAETYCK LHNSLIASIL DPYSNAFAWL QMETSNERVW IALNSNLTDN QYTWTDKWRV RYTNWAADEP KLKSACVYLD LDGYWKTAHC NESFYFLCK | |
Greater than 90% as determined by SDS-PAGE. |
4360 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |