missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lymphocyte antigen 6H Protein

A DNA sequence encoding the Homo sapiens (Human) Lymphocyte antigen 6H, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9259-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007941

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001123950.1
Lymphocyte antigen 6H Protein
Research Use Only
LY6H
Recombinant Protein
Yeast
LWCQDCTLTT NSSHCTPKQC QPSDTVCASV RITDPSSSRK DHSVNKMCAS SCDFVKRHFF SDYLMGFINS GILKVDVDCC EKDLCNGAAG
Greater than 90% as determined by SDS-PAGE.
4062
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers