missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lymphocyte antigen 6H Protein
A DNA sequence encoding the Homo sapiens (Human) Lymphocyte antigen 6H, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_001123950.1 |
---|---|
Gene ID (Entrez) | 4062 |
Name | Lymphocyte antigen 6H Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087931
|
enQuireBio™
QP9259-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027941
|
enQuireBio™
QP9259-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097931
|
enQuireBio™
QP9259-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017941
|
enQuireBio™
QP9259-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037941
|
enQuireBio™
QP9259-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007941
|
enQuireBio™
QP9259-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001123950.1 | |
Lymphocyte antigen 6H Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4062 | |
Research Use Only | |
LY6H | |
Recombinant Protein | |
Yeast | |
LWCQDCTLTT NSSHCTPKQC QPSDTVCASV RITDPSSSRK DHSVNKMCAS SCDFVKRHFF SDYLMGFINS GILKVDVDCC EKDLCNGAAG | |
Greater than 90% as determined by SDS-PAGE. |