missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LSM2 Protein

A cDNA sequence encoding the LSM2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12591-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960919

  • 3585.00 EUR / 1mg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

LSM2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMLFYSF FKSLVGKDVV VELKNDLSIC GTLHSVDQYL NIKLTDISVT DPEKYPHMLS VKNCFIRGSV VRYVQLPADE VDTQLLQDAA RKEALQQKQ
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LSM2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM Nacl, 5mM DTT and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers