missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lipolysis-stimulated lipoprotein receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Lipolysis-stimulated lipoprotein receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7800-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001247418.1 | |
Lipolysis-stimulated lipoprotein receptor Protein | |
Research Use Only | |
LSR | |
Recombinant Protein | |
E. coli | |
MQQDGLGVGT RNGSGKGRSV HPSWPWCAPR PLRYFGRDAR ARRAQTAAMA LLAGGLSRGL GSHPAAAGRD AVVFVWLLLS TWCTAPARAI QVTVSNPYHV VILFQPVTLP CTYQMTSTPT QPIVIWKYKS FCRDRIADAF SPASVDNQLN AQLAAGNPGY NPYVECQDSV RTVRVVATKQ GNAVTLGDYY QGRRITITGN ADLTFDQTAW GDSGVYYCSV VSAQDLQGNN EAYAELIVLG RTSGVAELLP GFQAGPIED | |
Greater than 90% as determined by SDS-PAGE. |
51599 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |