missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIM and SH3 domain protein 1

A DNA sequence encoding the Homo sapiens (Human) LIM and SH3 domain protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8479-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054841

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001258537.1
LIM and SH3 domain protein 1
Research Use Only
LASP1
Recombinant Protein
E. coli
MNPNCARCGK IVYPTEKVNC LDKFWHKACF HCETCKMTLN MKNYKGYEKK PYCNAHYPKQ SFTMVADTPE NLRLKQQSEL QSQVRYKEEF EKNKGKGFSV VADTPELQRI KKTQDQISNI KYHEEFEKSR MGPSGGEGME PERRDSQDGS SYRRPLEQQQ PHHIPTSAPV YQQPQQQPVA QSYGGYKEPA APVSIQRSAP GGGGKRYRAV YDYSAADEDE VSFQDGDTIV NVQQIDDGWM YGT
Greater than 90% as determined by SDS-PAGE.
3927
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers