missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LHPP Protein

A DNA sequence encoding the Homo sapiens (Human) LHPP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8086-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022581

  • 458.00 EUR / 100µg
Estimated Shipment: 03-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001161352.1
LHPP Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
64077
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LHPP
Recombinant Protein
E. coli
MAPWGKRLAG VRGVLLDISG VLYDSGAGGG TAIAGSVEAV ARLKRSRLKV RFCTNESQKS RAELVGQLQR LGFDISEQEV TAPAPAACQI LKEQGLRPYL LIHDGVRSEF DQIDTSNPNC VVIADAGESF SYQNMNNAFQ VLMELEKPVL ISLGKGRYYK ETSGLMLDVG PYMKALEYAC GIKAEVVGKP SPEFFKSALQ AIGVEAHQAV MIGDDIVGDV GGAQRCGMRA LQVRTGKFRP SDEHHPEVKA DGYVDNLAEA VDLLLQHADK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers