missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Laminin subunit beta-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Laminin subunit beta-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8788-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002283.3 | |
Laminin subunit beta-2 Protein | |
Research Use Only | |
LAMB2 | |
Recombinant Protein | |
E. coli | |
PDSIEMVATR VLELSIPASA EQIQHLAGAI AERVRSLADV DAILARTVGD VRRAEQLLQD ARRARSWAED EKQKAETVQA ALEEAQRAQG IAQGAIRGAV ADTRDTEQTL YQVQERMAGA ERALSSAGER ARQLDALLEA LKLKRAGNSL AASTAEETAG SAQGRAQEAE QLLRGPLGDQ YQTVKALAER KAQGVLAAQA RAEQLRDEAR DLLQAAQDKL QRLQELEGTY EENERALESK AAQLDGLEAR MRSVLQAINL QVQIYNTCQ | |
Greater than 90% as determined by SDS-PAGE. |
3913 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |