missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human L2HGDH, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) L2HGDH, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8068-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_079160.1 | |
L2HGDH, mitochondrial Protein | |
Research Use Only | |
L2HGDH | |
Recombinant Protein | |
E. coli | |
VIVGGGIVGL ASARALILRH PSLSIGVLEK EKDLAVHQTG HNSGVIHSGI YYKPESLKAK LCVQGAALLY EYCQQKGISY KQCGKLIVAV EQEEIPRLQA LYEKGLQNGV PGLRLIQQED IKKKEPYCRG LMAIDCPHTG IVDYRQVALS FAQDFQEAGG SVLTNFEVKG IEMAKESPSR SIDGMQYPIV IKNTKGEEIR CQYVVTCAGL YSDRISELSG CTPDPRIVPF RGDYLLLKPE KCYLVKGNIY PVPDSRFPFL GVHFTPRMDG SIWLGPNAVL AFKREGYRPF DFSATDVMDI IINSGLIKLA SQNFSYGVTE MYKACFLGAT VKYLQKFIPE ITISDILRGP AGVRAQALDR DGNLVEDFVF DAGVGDIGNR ILHVRNAPSP AATSSIAISG MIADEVQQRF EL | |
Greater than 90% as determined by SDS-PAGE. |
79944 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |