missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human L-lactate dehydrogenase C chain Protein
A DNA sequence encoding the Homo sapiens (Human) L-lactate dehydrogenase C chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6291-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002292.1 | |
L-lactate dehydrogenase C chain Protein | |
Research Use Only | |
LDHC | |
Recombinant Protein | |
E. coli | |
STVKEQLIEK LIEDDENSQC KITIVGTGAV GMACAISILL KDLADELALV DVALDKLKGE MMDLQHGSLF FSTSKITSGK DYSVSANSRI VIVTAGARQQ EGETRLALVQ RNVAIMKSII PAIVHYSPDC KILVVSNPVD ILTYIVWKIS GLPVTRVIGS GCNLDSARFR YLIGEKLGVH PTSCHGWIIG EHGDSSVPLW SGVNVAGVAL KTLDPKLGTD SDKEHWKNIH KQVIQSAYEI IKLKGYTSWA IGLSVMDLVG SILKNLRRVH PVSTMVKGLY GIKEELFLSI PCVLGRNGVS DVVKINLNSE EEALFKKSAE TLWNIQKDLI F | |
Greater than 90% as determined by SDS-PAGE. |
3948 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |