missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kita-kyushu lung cancer antigen 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Kita-kyushu lung cancer antigen 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7682-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020161

  • 675.00 EUR / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001017978.1
Kita-kyushu lung cancer antigen 1 Protein
Research Use Only
CT83
Recombinant Protein
Yeast
MNFYLLLASS ILCALIVFWK YRRFQRNTGE MSSNSTALAL VRPSSSGLIN SNTDNNLAVY DLSRDILNNF PHSIARQKRI LVNLSMVENK LVELEHTLLS KGFRGASPHR KST
Greater than 80% as determined by SDS-PAGE.
203413
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers