missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIR2DS1 Protein

A DNA sequence encoding the Homo sapiens (Human) KIR2DS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7614-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_055327.1
KIR2DS1 Protein
Research Use Only
KIR2DS1
Recombinant Protein
Yeast
HEGVHRKPSL LAHPGRLVKS EETVILQCWS DVMFEHFLLH REGMFNDTLR LIGEHHDGVS KANFSISRMR QDLAGTYRCY GSVTHSPYQL SAPSDPLDIV IIGLYEKPSL SAQPGPTVLA GENVTLSCSS RSSYDMYHLS REGEAHERRL PAGTKVNGTF QANFPLGPAT HGGTYRCFGS FRDSPYEWSK SSDPLLVSVT GNPSNSWPSP TEPSSETGNP RHLH
Greater than 90% as determined by SDS-PAGE.
3806
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers