missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIR2DS1 Protein
A DNA sequence encoding the Homo sapiens (Human) KIR2DS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7614-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055327.1 | |
KIR2DS1 Protein | |
Research Use Only | |
KIR2DS1 | |
Recombinant Protein | |
Yeast | |
HEGVHRKPSL LAHPGRLVKS EETVILQCWS DVMFEHFLLH REGMFNDTLR LIGEHHDGVS KANFSISRMR QDLAGTYRCY GSVTHSPYQL SAPSDPLDIV IIGLYEKPSL SAQPGPTVLA GENVTLSCSS RSSYDMYHLS REGEAHERRL PAGTKVNGTF QANFPLGPAT HGGTYRCFGS FRDSPYEWSK SSDPLLVSVT GNPSNSWPSP TEPSSETGNP RHLH | |
Greater than 90% as determined by SDS-PAGE. |
3806 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |