missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Keratin, type I cuticular Ha5 Protein

A DNA sequence encoding the Homo sapiens (Human) Keratin, type I cuticular Ha5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7995-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032071

  • 2325.00 EUR / 500µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002271.3
Keratin, type I cuticular Ha5 Protein
Research Use Only
KRT35
Recombinant Protein
Yeast
MASKCLKAGF SSGSLKSPGG ASGGSTRVSA MYSSSSCKLP SLSPVARSFS ACSVGLGRSS YRATSCLPAL CLPAGGFATS YSGGGGWFGE GILTGNEKET MQSLNDRLAG YLEKVRQLEQ ENASLESRIR EWCEQQVPYM CPDYQSYFRT IEELQKKTLC SKAENARLVV EIDNAKLAAD DFRTKYETEV SLRQLVESDI NGLRRILDDL TLCKSDLEAQ VESLKEELLC LKKNHEEEVN SLRCQLGDRL NVEVDAAPPV DLNRVLEEMR CQYETLVENN RRDAEDWLDT QSEELNQQVV SSSEQLQSCQ AEIIELRRTV NALEIELQAQ HSMRDALEST LAETEARYSS QLAQMQCMIT NVEAQLAEIR ADLERQNQEY QVLLDVRARL ECEINTYRGL LESEDSKLPC NPCAPDYSPS KSCLPCLPAA SCGPSAARTN CSPRPICVPC PGGRF
Greater than 90% as determined by SDS-PAGE.
3886
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers