missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kallikrein-10 Protein
A DNA sequence encoding the Homo sapiens (Human) Kallikrein-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8778-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001070968.1 | |
Kallikrein-10 Protein | |
Research Use Only | |
KLK10 | |
Recombinant Protein | |
E. coli | |
AEAALLPQND TRLDPEAYGS PCARGSQPWQ VSLFNGLSFH CAGVLVDQSW VLTAAHCGNK PLWARVGDDH LLLLQGEQLR RTTRSVVHPK YHQGSGPILP RRTDEHDLML LKLARPVVLG PRVRALQLPY RCAQPGDQCQ VAGWGTTAAR RVKYNKGLTC SSITILSPKE CEVFYPGVVT NNMICAGLDR GQDPCQSDSG GPLVCDETLQ GILSWGVYPC GSAQHPAVYT QICKYMSWIN KVIR | |
Greater than 90% as determined by SDS-PAGE. |
5655 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |