missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human JAM-2 / JAM-B Protein

A DNA sequence encoding the Homo sapiens (Human) JAM-2 / JAM-B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6247-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001257336.1
JAM-2 / JAM-B Protein
Research Use Only
JAM2
Recombinant Protein
Yeast
FSAPKDQQVV TAVEYQEAIL ACKTPKKTVS SRLEWKKLGR SVSFVYYQQT LQGDFKNRAE MIDFNIRIKN VTRSDAGKYR CEVSAPSEQG QNLEEDTVTL EVLVAPAVPS CEVPSSALSG TVVELRCQDK EGNPAPEYTW FKDGIRLLEN PRLGSQSTNS SYTMNTKTGT LQFNTVSKLD TGEYSCEARN SVGYRRCPGK RMQVDDLNIS
Greater than 90% as determined by SDS-PAGE.
58494
500 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions