missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JAM-2 / JAM-B Protein
A DNA sequence encoding the Homo sapiens (Human) JAM-2 / JAM-B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6247-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257336.1 | |
JAM-2 / JAM-B Protein | |
Research Use Only | |
JAM2 | |
Recombinant Protein | |
Yeast | |
FSAPKDQQVV TAVEYQEAIL ACKTPKKTVS SRLEWKKLGR SVSFVYYQQT LQGDFKNRAE MIDFNIRIKN VTRSDAGKYR CEVSAPSEQG QNLEEDTVTL EVLVAPAVPS CEVPSSALSG TVVELRCQDK EGNPAPEYTW FKDGIRLLEN PRLGSQSTNS SYTMNTKTGT LQFNTVSKLD TGEYSCEARN SVGYRRCPGK RMQVDDLNIS | |
Greater than 90% as determined by SDS-PAGE. |
58494 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |