missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Islet cell autoantigen 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Islet cell autoantigen 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6188-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031430

  • 2090.00 EUR / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129492.1
Islet cell autoantigen 1 Protein
Research Use Only
ICA1
Recombinant Protein
E. coli
MSGHKCSYPW DLQDRYAQDK SVVNKMQQKY WETKQAFIKA TGKKEDEHVV ASDADLDAKL ELFHSIQRTC LDLSKAIVLY QKRICFLSQE ENELGKFLRS QGFQDKTRAG KMMQATGKAL CFSSQQRLAL RNPLCRFHQE VETFRHRAIS DTWLTVNRME QCRTEYRGAL LWMKDVSQEL DPDLYKQMEK FRKVQTQVRL AKKNFDKLKM DVCQKVDLLG ASRCNLLSHM LATYQTTLLH FWEKTSHTMA AIHESFKGYQ PYEFTTLK
Greater than 90% as determined by SDS-PAGE.
3382
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers