missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin alpha-V Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-V, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_001138471.1 |
---|---|
Gene ID (Entrez) | 3685 |
Name | Integrin alpha-V Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017871
|
enQuireBio™
QP9224-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057871
|
enQuireBio™
QP9224-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027871
|
enQuireBio™
QP9224-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047871
|
enQuireBio™
QP9224-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067871
|
enQuireBio™
QP9224-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037871
|
enQuireBio™
QP9224-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001138471.1 | |
Integrin alpha-V Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3685 | |
Research Use Only | |
ITGAV | |
Recombinant Protein | |
Yeast | |
DLALSEGDIH TLGCGVAQCL KIVCQVGRLD RGKSAILYVK SLLWTETFMN KENQNHSYSL KSSASFNVIE FPYKNLPIED ITNSTLVTTN VTWGIQPAPM PVPVWVIILA VLAGLLLLAV LVFVMYRMGF FKRVRPPQEE QEREQLQPHE NGEGNSET | |
Greater than 90% as determined by SDS-PAGE. |