missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin alpha-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6238-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031830

  • 565.00 EUR / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002194.2
Integrin alpha-2 Protein
Research Use Only
ITGA2
Recombinant Protein
Yeast
WDAVKNFLEK FVQGLDIGPT KTQVGLIQYA NNPRVVFNLN TYKTKEEMIV ATSQTSQYGG DLTNTFGAIQ YARKYAYSAA SGGRRSATKV MVVVTDGESH DGSMLKAVID QCNHDNILRF GIAVLGYLNR NALDTKNLIK EIKAIASIPT ERYFFNVSDE AALLEKAGTL GEQIFSIE
Greater than 90% as determined by SDS-PAGE.
3673
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers