missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inactive phospholipase D5 Protein
A DNA sequence encoding the Homo sapiens (Human) Inactive phospholipase D5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7881-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001182740.1 | |
Inactive phospholipase D5 Protein | |
Research Use Only | |
PLD5 | |
Recombinant Protein | |
E. coli | |
MGEDEDGLSE KNCQNKCRIA LVENIPEGLN YSENAPFHLS LFQGWMNLLN MAKKSVDIVS SHWDLNHTHP SACQGQRLFE KLLQLTSQNI EIKLVSDVTA DSKVLEALKL KGAEVTYMNM TAYNKGRLQS SFWIVDKQHV YIGSAGLDWQ SLGQMKELGV IFYNCSCLVL DLQRIFALYS SLKFKSRVPQ TWSKRLYGVY DNEKKLQLQL NETKSQAFVS NSPKLFCPKN RSFDIDAIYS VIDDAKQYVY IAVMDYLPIS STSTKRTYWP DLDAKIREAL VLRSVRVRLL LSFWKETDPL TFNFISSLKA ICTEIANCSL KVKFFDLERE NACATKEQKN HTFPRLNRNK YMVTDGAAYI GNFDWVGNDF TQNAGTGLVI NQADVRNNRS IIKQLKDVFE RDWYSPYAKT LQPTKQPNCS SLFKLKPLSN KTATDDTGGK DPRNV | |
Greater than 90% as determined by SDS-PAGE. |
200150 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |