missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Immunoglobulin iota chain Protein
A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin iota chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6882-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009059.1 | |
Immunoglobulin iota chain Protein | |
Research Use Only | |
VPREB1 | |
Recombinant Protein | |
E. coli | |
QPVLHQPPAM SSALGTTIRL TCTLRNDHDI GVYSVYWYQQ RPGHPPRFLL RYFSQSDKSQ GPQVPPRFSG SKDVARNRGY LSISELQPED EAMYYCAMGA RSSEKEERER EWEEEMEPTA ARTRVP | |
Greater than 90% as determined by SDS-PAGE. |
7441 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |