missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL36G / IL1F9 Protein
A DNA sequence encoding the Homo sapiens (Human) IL36G / IL1F9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8125-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265497.1 | |
IL36G / IL1F9 Protein | |
Research Use Only | |
IL36G | |
Recombinant Protein | |
E. coli | |
MRGTPGDADG GGRAVYQSMC KPITGTINDL NQQVWTLQGQ NLVAVPRSDS VTPVTVAVIT CKYPEALEQG RGDPIYLGIQ NPEMCLYCEK VGEQPTLQLK EQKIMDLYGQ PEPVKPFLFY RAKTGRTSTL ESVAFPDWFI ASSKRDQPII LTSELGKSYN TAFELNIND | |
Greater than 80% as determined by SDS-PAGE. |
56300 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |