missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL1F6 / IL36 Protein

A DNA sequence encoding the Homo sapiens (Human) IL1F6 / IL36, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8208-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093281

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_055255.1
IL1F6 / IL36 Protein
Research Use Only
Il36a
Recombinant Protein
E. coli
MEKALKIDTP QQGSIQDINH RVWVLQDQTL IAVPRKDRMS PVTIALISCR HVETLEKDRG NPIYLGLNGL NLCLMCAKVG DQPTLQLKEK DIMDLYNQPE PVKSFLFYHS QSGRNSTFES VAFPGWFIAV SSEGGCPLIL TQELGKANTT DFGLTMLF
Greater than 80% as determined by SDS-PAGE.
27179
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers