missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F6 / IL36 Protein
A DNA sequence encoding the Homo sapiens (Human) IL1F6 / IL36, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8208-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055255.1 | |
IL1F6 / IL36 Protein | |
Research Use Only | |
Il36a | |
Recombinant Protein | |
E. coli | |
MEKALKIDTP QQGSIQDINH RVWVLQDQTL IAVPRKDRMS PVTIALISCR HVETLEKDRG NPIYLGLNGL NLCLMCAKVG DQPTLQLKEK DIMDLYNQPE PVKSFLFYHS QSGRNSTFES VAFPGWFIAV SSEGGCPLIL TQELGKANTT DFGLTMLF | |
Greater than 80% as determined by SDS-PAGE. |
27179 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |