missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL18BP Protein
A DNA sequence encoding the Homo sapiens (Human) IL18BP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8878-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034748.1 | |
IL18BP Protein | |
Research Use Only | |
IL18BP | |
Recombinant Protein | |
E. coli | |
TPVSQTTTAA TASVRSTKDP CPSQPPVFPA AKQCPALEVT WPEVEVPLNG TLSLSCVACS RFPNFSILYW LGNGSFIEHL PGRLWEGSTS RERGSTGTQL CKALVLEQLT PALHSTNFSC VLVDPEQVVQ RHVVLAQLWA GLRATLPPTQ EAL | |
Greater than 90% as determined by SDS-PAGE. |
10068 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |