missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein

A DNA sequence encoding the Homo sapiens (Human) IL-29 / Interleukin-29, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7814-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030951

  • 463.00 EUR / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_742152.1
IL-29 / Interleukin-29 Protein
Research Use Only
IFNL1
Recombinant Protein
E. coli
PVPTSKPTTT GKGCHIGRFK SLSPQELASF KKARDALEES LKLKNWSCSS PVFPGNWDLR LLQVRERPVA LEAELALTLK VLEAAAGPAL EDVLDQPLHT LHHILSQLQA CIQPQPTAGP RPRGRLHHWL HRLQEAPKKE SAGCLEASVT FNLFRLLTRD LKYVADGNLC LRTSTHPEST
Greater than 90% as determined by SDS-PAGE.
282618
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers