missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGF-2 / IGF-II Protein

A DNA sequence encoding the Homo sapiens (Human) IGF-2 / IGF-II, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8601-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035511

  • 463.00 EUR / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000603.1
IGF-2 / IGF-II Protein
Research Use Only
IGF2
Recombinant Protein
E. coli
ETLCGGELVD TLQFVCGDRG FYFSRPASRV SRRSRGIVEE CCFRSCDLAL LETYCATPAK SERDVSTPPT VLPDNFPRYP VGKFFQYDTW KQSTQRLRRG LPALLRARRG HVLAKELEAF REAKRHRPLI ALPTQDPAHG GAPPEMASNR K
Greater than 90% as determined by SDS-PAGE.
3481
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers