missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSD17B14 Protein

A DNA sequence encoding the Homo sapiens (Human) HSD17B14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8019-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_057330.2
HSD17B14 Protein
Research Use Only
HSD17B14
Recombinant Protein
E. coli
MATGTRYAGK VVVVTGGGRG IGAGIVRAFV NSGARVVICD KDESGGRALE QELPGAVFIL CDVTQEDDVK TLVSETIRRF GRLDCVVNNA GHHPPPQRPE ETSAQGFRQL LELNLLGTYT LTKLALPYLR KSQGNVINIS SLVGAIGQAQ AVPYVATKGA VTAMTKALAL DESPYGVRVN CISPGNIWTP LWEELAALMP DPRATIREGM LAQPLGRMGQ PAEVGAAAVF LASEANFCTG IELLVTGGAE LGYGCKASRS TPVDAPDIPS
Greater than 90% as determined by SDS-PAGE.
51171
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers