Learn More
enQuireBio™ Recombinant Human HSD17B14 Protein
A DNA sequence encoding the Homo sapiens (Human) HSD17B14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8019-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057330.2 | |
HSD17B14 Protein | |
Research Use Only | |
HSD17B14 | |
Recombinant Protein | |
E. coli | |
MATGTRYAGK VVVVTGGGRG IGAGIVRAFV NSGARVVICD KDESGGRALE QELPGAVFIL CDVTQEDDVK TLVSETIRRF GRLDCVVNNA GHHPPPQRPE ETSAQGFRQL LELNLLGTYT LTKLALPYLR KSQGNVINIS SLVGAIGQAQ AVPYVATKGA VTAMTKALAL DESPYGVRVN CISPGNIWTP LWEELAALMP DPRATIREGM LAQPLGRMGQ PAEVGAAAVF LASEANFCTG IELLVTGGAE LGYGCKASRS TPVDAPDIPS | |
Greater than 90% as determined by SDS-PAGE. |
51171 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |