missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSD11B2 Protein
A DNA sequence encoding the Homo sapiens (Human) HSD11B2, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_000187.3 |
---|---|
Gene ID (Entrez) | 3291 |
Name | HSD11B2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057801
|
enQuireBio™
QP9185-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097801
|
enQuireBio™
QP9185-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067801
|
enQuireBio™
QP9185-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087801
|
enQuireBio™
QP9185-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007811
|
enQuireBio™
QP9185-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077801
|
enQuireBio™
QP9185-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000187.3 | |
HSD11B2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3291 | |
Research Use Only | |
HSD11B2 | |
Recombinant Protein | |
Yeast | |
MERWPWPSGG AWLLVAARAL LQLLRSDLRL GRPLLAALAL LAALDWLCQR LLPPPAALAV LAAAGWIALS RLARPQRLPV ATRAVLITGC DSGFGKETAK KLDSMGFTVL ATVLELNSPG AIELRTCCSP RLRLLQMDLT KPGDISRVLE FTKAHTTSTG LWGLVNNAGH NEVVADAELS PVATFRSCME VNFFGALELT KGLLPLLRSS RGRIVTVGSP AGDMPYPCLG AYGTSKAAVA LLMDTFSCEL LPWGVKVSII QPGCFKTESV RNVGQWEKRK QLLLANLPQE LLQAYGKDYI EHLHGQFLHS LRLAMSDLTP VVDAITDALL AARPRRRYYP GQGLGLMYFI HYYLPEGLRR RFLQAFFISH CLPRALQPGQ PGTTPPQDAA QDPNLSPGPS PAVAR | |
Greater than 90% as determined by SDS-PAGE. |