missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HPRT1 Protein
A DNA sequence encoding the Homo sapiens (Human) HPRT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8846-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000185.1 | |
HPRT1 Protein | |
Research Use Only | |
HPRT1 | |
Recombinant Protein | |
E. coli | |
ATRSPGVVIS DDEPGYDLDL FCIPNHYAED LERVFIPHGL IMDRTERLAR DVMKEMGGHH IVALCVLKGG YKFFADLLDY IKALNRNSDR SIPMTVDFIR LKSYCNDQST GDIKVIGGDD LSTLTGKNVL IVEDIIDTGK TMQTLLSLVR QYNPKMVKVA SLLVKRTPRS VGYKPDFVGF EIPDKFVVGY ALDYNEYFRD LNHVCVISET GKAKYKA | |
Greater than 90% as determined by SDS-PAGE. |
3251 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |