missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HPRT1 Protein

A DNA sequence encoding the Homo sapiens (Human) HPRT1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8846-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000185.1
HPRT1 Protein
Research Use Only
HPRT1
Recombinant Protein
E. coli
ATRSPGVVIS DDEPGYDLDL FCIPNHYAED LERVFIPHGL IMDRTERLAR DVMKEMGGHH IVALCVLKGG YKFFADLLDY IKALNRNSDR SIPMTVDFIR LKSYCNDQST GDIKVIGGDD LSTLTGKNVL IVEDIIDTGK TMQTLLSLVR QYNPKMVKVA SLLVKRTPRS VGYKPDFVGF EIPDKFVVGY ALDYNEYFRD LNHVCVISET GKAKYKA
Greater than 90% as determined by SDS-PAGE.
3251
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers