missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein OTX2 Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein OTX2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001257452.1 |
---|---|
Gene ID (Entrez) | 5015 |
Name | Homeobox protein OTX2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043440
|
enQuireBio™
QP6458-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063440
|
enQuireBio™
QP6458-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053440
|
enQuireBio™
QP6458-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073440
|
enQuireBio™
QP6458-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083440
|
enQuireBio™
QP6458-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093440
|
enQuireBio™
QP6458-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257452.1 | |
Homeobox protein OTX2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5015 | |
Research Use Only | |
OTX2 | |
Recombinant Protein | |
E. coli | |
MMSYLKQPPY AVNGLSLTTS GMDLLHPSVG YPGPWASCPA ATPRKQRRER TTFTRAQLDV LEALFAKTRY PDIFMREEVA LKINLPESRV QVWFKNRRAK CRQQQQQQQN GGQNKVRPAK KKTSPAREVS SESGTSGQFT PPSSTSVPTI ASSSAPVSIW SPASISPLSD PLSTSSSCMQ RSYPMTYTQA SGYSQGYAGS TSYFGGMDCG SYLTPMHHQL PGPGATLSPM GTNAVTSHLN QSPASLSTQG YGASSLGFNS TTDCLDYKDQ TASWKLNFNA DCLDYKDQTS SWKFQVL | |
Greater than 90% as determined by SDS-PAGE. |