missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DRB Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DRB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8911-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
HLA-DRB Protein | |
Research Use Only | |
HLA-DRB1 | |
Recombinant Protein | |
E. coli | |
DTRPRFLEYS TGECYFFNGT ERVRLLERHF HNQEELLRFD SDVGEFRAVT ELGRPVAESW NSQKDILEDR RAAVDTYCRH NYGAVESFTV QRRVHPKVTV YPSKTQPLQH HNLLVCSVSG FYPGSIEVRW FRNGQEEKTG VVSTGLIHNG DWTFQTLVML ETVPRSGEVY TCQVEHPSVT SPLTVEWRAR SESAQSKMLS GVGGFVLGLL FLGAGLFIYF RNQKGHSGLQ PRGFLS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |