missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DQA1 Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DQA1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8817-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006651

  • 1855.00 EUR / 1mg


Estimated Shipment
23-03-2020
Add to basket

Specifications

Specifications

NP_002113.2
HLA-DQA1 Protein
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
3117
Greater than 80% as determined by SDS-PAGE.
Research Use Only
HLA-DQA1
Recombinant Protein
E. coli
EDIVADHVAS YGVNLYQSYG PSGQYTHEFD GDEQFYVDLG RKETVWCLPV LRQFRFDPQF ALTNIAVLKH NLNSLIKRSN STAATNEVPE VTVFSKSPVT LGQPNILICL VDNIFPPVVN ITWLSNGHSV TEGVSETSFL SKSDHSFFKI SYLTLLPSAE ESYDCKVEHW GLDKPLLKHW EPEIPAPMSE
Videos
Documents