missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DPA1 Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DPA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8815-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001229453.1 | |
HLA-DPA1 Protein | |
Research Use Only | |
HLA-DPA1 | |
Recombinant Protein | |
E. coli | |
AGAIKADHVS TYAAFVQTHR PTGEFMFEFD EDEMFYVDLD KKETVWHLEE FGQAFSFEAQ GGLANIAILN NNLNTLIQRS NHTQATNDPP EVTVFPKEPV ELGQPNTLIC HIDKFFPPVL NVTWLCNGEL VTEGVAESLF LPRTDYSFHK FHYLTFVPSA EDFYDCRVEH WGLDQPLLKH WEAQEPIQMP ETTE | |
Greater than 90% as determined by SDS-PAGE. |
3113 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |