missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DOB Protein

A cDNA sequence encoding the HLA-DOB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12275-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959908

  • 120.00 EUR / 5µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

HLA-DOB Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGTDSPED FVIQAKADCY FTNGTEKVQF VVRFIFNLEE YVRFDSDVGM FVALTKLGQP DAEQWNSRLD LLERSRQAVD GVCRHNYRLG APFTVGRKVQ PEVTVYPERT PLLHQHNLLH CSVTGFYPGD IKIKWFLNGQ EERAGVMSTG PIRNGDWTFQ TVVMLEMTPE LGHVYTCLVD HSSLLSPVSV EWRAQSEYSW RK
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HLA-DOB solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers