missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DMA Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DMA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7191-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006111.2 | |
HLA-DMA Protein | |
Research Use Only | |
HLA-DMA | |
Recombinant Protein | |
E. coli | |
VPEAPTPMWP DDLQNHTFLH TVYCQDGSPS VGLSEAYDED QLFFFDFSQN TRVPRLPEFA DWAQEQGDAP AILFDKEFCE WMIQQIGPKL DGKIPVSRGF PIAEVFTLKP LEFGKPNTLV CFVSNLFPPM LTVNWQHHSV PVEGFGPTFV SAVDGLSFQA FSYLNFTPEP SDIFSCIVTH EIDRYTAIAY WVPRNALPSD LLENVLC | |
Greater than 90% as determined by SDS-PAGE. |
3108 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |