missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-C Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-C, was expressed in the hosts and tags indicated.
160.00 EUR - 1180.00 EUR
Specifications
Name | HLA-C Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HLA-C |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046681
|
enQuireBio™
QP8825-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086681
|
enQuireBio™
QP8825-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056681
|
enQuireBio™
QP8825-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066681
|
enQuireBio™
QP8825-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076681
|
enQuireBio™
QP8825-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
HLA-C Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HLA-C | |
Recombinant Protein | |
E. coli | |
CSHSMRYFYT AVSRPGRGEP RFIAVGYVDD TQFVRFDSDA ASPRGEPRAP WVEQEGPEYW DRETQKYKRQ AQTDRVSLRN LRGYYNQSEA GSHTLQWMYG CDLGPDGRLL RGYDQSAYDG KDYIALNEDL RSWTAADTAA QITQRKWEAA RAAEQQRAYL EGTCVEWLRR YLENGKETLQ RAEHPKTHVT HHLVSDHEAT LRCWALGFYP AEITLTWQRD GEDQTQDTEL VETRPAGDGT FQKWAAVVVP SGEEQRYTCH VQHEGLPEPL TLRWEPSSQP TIPI | |
Greater than 90% as determined by SDS-PAGE. |