missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-C Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8825-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
HLA-C Protein | |
Research Use Only | |
HLA-C | |
Recombinant Protein | |
E. coli | |
CSHSMRYFYT AVSRPGRGEP RFIAVGYVDD TQFVRFDSDA ASPRGEPRAP WVEQEGPEYW DRETQKYKRQ AQTDRVSLRN LRGYYNQSEA GSHTLQWMYG CDLGPDGRLL RGYDQSAYDG KDYIALNEDL RSWTAADTAA QITQRKWEAA RAAEQQRAYL EGTCVEWLRR YLENGKETLQ RAEHPKTHVT HHLVSDHEAT LRCWALGFYP AEITLTWQRD GEDQTQDTEL VETRPAGDGT FQKWAAVVVP SGEEQRYTCH VQHEGLPEPL TLRWEPSSQP TIPI | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |