missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 11, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001129513.1 |
---|---|
Gene ID (Entrez) | 79885 |
Name | Histone deacetylase 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011190
|
enQuireBio™
QP6151-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031190
|
enQuireBio™
QP6151-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021190
|
enQuireBio™
QP6151-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041190
|
enQuireBio™
QP6151-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051190
|
enQuireBio™
QP6151-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061190
|
enQuireBio™
QP6151-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001129513.1 | |
Histone deacetylase 11 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79885 | |
Research Use Only | |
HDAC11 | |
Recombinant Protein | |
E. coli | |
MLHTTQLYQH VPETRWPIVY SPRYNITFMG LEKLHPFDAG KWGKVINFLK EEKLLSDSML VEAREASEED LLVVHTRRYL NELKWSFAVA TITEIPPVIF LPNFLVQRKV LRPLRTQTGG TIMAGKLAVE RGWAINVGGG FHHCSSDRGG GFCAYADITL AIKFLFERVE GISRATIIDL DAHQGNGHER DFMDDKRVYI MDVYNRHIYP GDRFAKQAIR RKVELEWGTE DDEYLDKVER NIKKSLQEHL PDVVVYNAGT DILEGDRLGG LSISPAGIVK RDELVFRMVR GRRVPILMVT SGGYQKRTAR IIADSILNLF GLGLIGPESP SVSAQNSDTP LLPPAVP | |
Greater than 80% as determined by SDS-PAGE. |