missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone acetyltransferase KAT7 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone acetyltransferase KAT7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8459-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024741

  • 463.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001186084.1
Histone acetyltransferase KAT7 Protein
Research Use Only
HBO1
Recombinant Protein
E. coli
MPRRKRNAGS SSDGTEDSDF STDLEHTDSS ESDGTSRRSA RVTRSSARLS QSSQDSSPVR NLQSFGTEEP AYSTRRVTRS QQQPTPVTPK KYPLRQTRSS GSETEQVVDF SDRETKNTAD HDESPPRTPT GNAPSSESDI DISSPNVSHD ESIAKDMSLK DSGSDLSHRP KRRRFHESYN FNMKCPTPGC NSLGHLTGKH ERHFSISGCP LYHN
Greater than 90% as determined by SDS-PAGE.
11143
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers