missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histidine decarboxylase Protein

A DNA sequence encoding the Homo sapiens (Human) Histidine decarboxylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9176-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007781

  • 416.00 EUR / 50µg


Estimated Shipment
21-02-2020
Add to basket

Specifications

Specifications

NP_001293075.1
Histidine decarboxylase Protein
50 μg
Not determined.
Full. Length
Human
His
Tris-based buffer, 50% glycerol
3067
Greater than 80% as determined by SDS-PAGE.
Research Use Only
HDC
Recombinant Protein
Yeast
MMEPEEYRER GREMVDYICQ YLSTVRERRV MPDVQPGYLR AQLPESAPED PDSWDSIFGD IERIIMPGVV HWQSPHMHAY YPALTSWPSL LGDMLADAIN CLGFTWASSP ACTELEMNVM DWLAKMLGLP EHFLHHHPSS QGGGVLQSTV SESTLIALLA ARKNKILEMK TSEPDADESC LNARLVAYAS DQAHSSVEKA GLISLVKMKF LPVDDNFSLR GEALQKAIEE DKQRGLVPVF VCATLGTTGV CAFDCLSELG PICAREGLWL HIDAAYAGTA FLCPEFRGFL KGIEYADSFT FNPSKWMMVH FDCTGFWVKD KYKLQQTFSV NPIYLRHANS GVATDFMHWQ IPLSRRFRSV KLWFVIRSFG VKNLQAHVRH GTEMAKYFES LVRNDPSFEI PAKRHLGLVV FRLKGPNCLT ENVLKEIAKA GRLFLIPATI QDKLIIRFTV TSQFTTRDDI LRDWNLIRDA ATLILSQHCT SQPSPRVGNL ISQIRGARAW ACGTSLQSVS GAGDDPVQAR KIIKQPQRVG AGPMKRENGL HLETLLDPVD DCFSEEAPDA TKHKLSSFLF SYLSVQTKKK TVRSLSCNSV PVSAQKPLPT EASVKNGGSS RVRIFSRFPE DMMMLKKSAF KKLIKFYSVP SFPECSSQCG LQLPCCPLQA MV
Videos
Documents