missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hemoglobin subunit zeta Protein

A DNA sequence encoding the Homo sapiens (Human) Hemoglobin subunit zeta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6148-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031170

  • 463.00 EUR / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005323.1
Hemoglobin subunit zeta Protein
Research Use Only
HBZ
Recombinant Protein
E. coli
SLTKTERTII VSMWAKISTQ ADTIGTETLE RLFLSHPQTK TYFPHFDLHP GSAQLRAHGS KVVAAVGDAV KSIDDIGGAL SKLSELHAYI LRVDPVNFKL LSHCLLVTLA ARFPADFTAE AHAAWDKFLS VVSSVLTEKY R
Greater than 80% as determined by SDS-PAGE.
3050
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers