missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock 70 kDa protein 13 Protein
A DNA sequence encoding the Homo sapiens (Human) Heat shock 70 kDa protein 13, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_008879.3 |
---|---|
Gene ID (Entrez) | 6782 |
Name | Heat shock 70 kDa protein 13 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006061
|
enQuireBio™
QP8708-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026061
|
enQuireBio™
QP8708-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016061
|
enQuireBio™
QP8708-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036061
|
enQuireBio™
QP8708-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046061
|
enQuireBio™
QP8708-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056061
|
enQuireBio™
QP8708-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_008879.3 | |
Heat shock 70 kDa protein 13 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6782 | |
Research Use Only | |
HSPA13 | |
Recombinant Protein | |
E. coli | |
QQYLPLPTPK VIGIDLGTTY CSVGVFFPGT GKVKVIPDEN GHISIPSMVS FTDNDVYVGY ESVELADSNP QNTIYDAKRF IGKIFTAEEL EAEIGRYPFK VLNKNGMVEF SVTSNETITV SPEYVGSRLL LKLKEMAEAY LGMPVANAVI SVPAEFDLKQ RNSTIEAANL AGLKILRVIN EPTAAAMAYG LHKADVFHVL VIDLGGGTLD VSLLNKQGGM FLTRAMSGNN KLGGQDFNQR LLQYLYKQIY QTYGFVPSRK EEIHRLRQAV EMVKLNLTLH QSAQLSVLLT VEEQDRKEPH SSDTELPKDK LSSADDHRVN SGFGRGLSDK KSGESQVLFE TEISRKLFDT LNEDLFQKIL VPIQQVLKEG HLEKTEIDEV VLVGGSTRIP RIRQVIQEFF GKDPNTSVDP DLAVVTGVAI QAGIDGGSWP LQVSALEIPN KHLQKTNFN | |
Greater than 90% as determined by SDS-PAGE. |