missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDAC4 Protein
A DNA sequence encoding the Homo sapiens (Human) HDAC4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8906-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_006028.2 | |
HDAC4 Protein | |
Research Use Only | |
HDAC4 | |
Recombinant Protein | |
E. coli | |
MSSQSHPDGL SGRDQPVELL NPARVNHMPS TVDVATALPL QVAPSAVPMD LRLDHQFSLP VAEPALREQQ LQQELLALKQ KQQIQRQILI AEFQRQHEQL SRQHEAQLHE HIKQQQEMLA MKHQQELLEH QRKLERHRQE QELEKQHREQ KLQQLKNKEK GKESAVASTE VKMKLQEFVL NKKKALAHRN LNHCISSDPR YWYGKTQHSS LDQSSPPQSG VSTSYNH | |
Greater than 90% as determined by SDS-PAGE. |
9759 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |