missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HAUS augmin-like complex subunit 4 Protein

A DNA sequence encoding the Homo sapiens (Human) HAUS augmin-like complex subunit 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8087-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042591

  • 575.00 EUR / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001159741.1
HAUS augmin-like complex subunit 4 Protein
Research Use Only
HAUS4
Recombinant Protein
E. coli
MASGDFCSPG EGMEILQQVC SKQLPPCNLS KEDLLQNPYF SKLLLNLSQH VDESGLSLTL AKEQAQAWKE VRLHKTTWLR SEILHRVIQE LLVDYYVKIQ DTNVTSEDKK FHETLEQRLL VTELMRLLGP SQEREIPPLL GLEKADLLEL MPLSEDFVWM RARLQQEVEE QLKKKCFTLL CYYDPNSDAD SETVKAAKVW KLAEVLVGEQ QQCQDAKSQQ KEQMLLLEKK SAAYSQVLLR CLTLLQRLLQ EHRLKTQSEL DRINAQYLEV KCGAMILKLR MEELKILSDT YTVEKVEVHR LIRDRLEGAI HLQEQDMENS RQVLNSYEVL GEEFDRLVKE YTVLKQATEN KRWALQEFSK VYR
Greater than 80% as determined by SDS-PAGE.
54930
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers