missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSTT1 Protein

A cDNA sequence encoding the GSTT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12064-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959248

  • 186.00 EUR / 20µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

GSTT1 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMGL ELYLDLLSQP CRAVYIFAKK NDIPFELRIV DLIKGQHLSD ACAQVNPLKK VPALKDGDFT LTESVAILLY LTRKYKVPDY WYPQDLQARA RVDEYLAWQH TTLRRSCLRA LWHKVMFPVF LGEPVSPQTL AATLAELDVT LQLLEDKFLQ NKAFLTGPHI SLADLVAITE LMHPVGAGCQ VFEGRPKLAT WRQRVEAAVG EDLFQEAHEV ILKAKDFPPA DPTIKQKLMP WVLAMIR
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GSTT1 solution contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers