missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNA15 Protein

A DNA sequence encoding the Homo sapiens (Human) GNA15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6100-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030800

  • 2985.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002059.3
GNA15 Protein
Research Use Only
GNA15
Recombinant Protein
E. coli
MARSLTWRCC PWCLTEDEKA AARVDQEINR ILLEQKKQDR GELKLLLLGP GESGKSTFIK QMRIIHGAGY SEEERKGFRP LVYQNIFVSM RAMIEAMERL QIPFSRPESK HHASLVMSQD PYKVTTFEKR YAAAMQWLWR DAGIRAYYER RREFHLLDSA VYYLSHLERI TEEGYVPTAQ DVLRSRMPTT GINEYCFSVQ KTNLRIVDVG GQKSERKKWI HCFENVIALI YLASLSEYDQ CLEENNQENR MKESLALFGT ILELPWFKST SVILFLNKTD ILEEKIPTSH LATYFPSFQG PKQDAEAAKR FILDMYTRMY TGCVDGPEGS KKGARSRRLF SHYTCATDTQ NIRKVFKDVR DSVLARYLDE INLL
Greater than 80% as determined by SDS-PAGE.
2769
1 mg
Not determined.
Full Length
Human
His-Trx
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers